Order Entry
Ireland
Orders LinkContactUsLinkComponent
Sampling swabs
Sampling swabs
PP/PE
Catalog # PMPC25-806-1PD
Supplier:   PURITAN MEDICAL
Sampling swabs
Catalog # PMPC25-806-1PD
Supplier:  PURITAN MEDICAL
Sampling swabs
Sampling swabs
PP/PE
Price
Inquire For Price
Log in for availability
1 * 1.000 items
Product Details & Documents
This synthetic tipped applicator is made of medical-grade polyester fibre that is carded in our factory to maintain consistency.
  • Standard tip with medical grade quality
  • Non-absorbent no fatty acids or latent sugars for specimen transport, lab work, and diagnostic kits
  • Bonded securely to a semi-flexible PS handle by an aqueous based adhesive
Applications include diagnostic specimen collection, patient care, cell collection for DNA testing and gynaecological screening.
Packaging: 10 boxes per case of 100 individually wrapped packages.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Specifications
  • Shaft/head:
    PP/PE