

Urea 98.0-102.0% USP, J.T.Baker®
: Avantor
:57-13-6
:
:NH₂CONH₂
:MFCD00008022
:13,09935
:197 °C (1013 hPa)
:132,7 °C
:1,34 g/cm³ (20 °C)
:60.06 g/mol
:Ambient
:200-315-5
:01-2119463277-33
:GHS07
:GHS08
- GMP Manufactured Product:Passes Test
- Meets U.S.P Requirements:Passes Test
- CAUTION: For Manufacturing, processing or repackaging:98.0 - 102.0%
- Bulk Pharmaceutical Chemical:≤0.1%
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Assay (NH₂CONH₂):98.0 - 102.0 %
- USP - Residue on Ignition:≤0.1 %
- USP - Alcohol Insoluble Matter:≤0.04 %
- USP - Organic impurities: Urea related compound A:≤0.1 %
- USP - Organic impurities: Any individual unspecified impurity:≤0.1 %
- USP - Organic impurities: Total impurities:≤2.0 %
Frequently Bought Together