Order Entry
Ireland
ContactUsLinkComponent
Sodium dihydrogen phosphate monohydrate Ph. Eur., Macron Fine Chemicals™
Sodium dihydrogen phosphate monohydrate Ph. Eur., Macron Fine Chemicals™
Catalog # P117-38
CAS Number:  
undefined
Sodium dihydrogen phosphate monohydrate Ph. Eur., Macron Fine Chemicals™
Catalog # P117-38
CAS Number:  

Specifications

  • Pk:
    25 kg
  • Seq:
    4283

Specifications

About this item

 :
 :137.99 g/mol
Merck Index:13,08734
Melting Pt:100 °C
Density:2,04 g/cm³
Storage Temperature:Ambient
EINECS:231-449-2
REACH:01-2119489796-13
CAS Number:10049-21-5
Formula:NaH₂PO₄·H₂O
MDL Number:MFCD00149208