

Potassium hydroxide ≥85.0%, pellets, GenAR® NF, Macron Fine Chemicals™
Catalog # 7815-88
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pack type:PE pail, screw top
- Pk:12 kg
- Seq:4187
Specifications
Specification Test Results
- Bulk Pharmaceutical Chemical:= 85.0 %
- Bulk Food Chemical:= 3.5 %
- NF - Assay (Total Alkali):≥85.0 %
- NF - Carbonate (as K₂CO₃):≤3.5 %
- NF - Identification A:Passes Test
- NF - Identification B:Passes Test
- NF - Insoluble Substances:Passes Test
- NF - Content of Potassium:≥59.9 %
- NF - Limit of Sodium:≤1.0 %
- Iron (Fe):≤0.0003 %
- EP/BP - Assay (total alkali as KOH):85.0 - 100.5 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Carbonate (as K₂CO₃):≤2.0 %
- EP/BP - Chloride (Cl):≤200 ppm
- EP/BP - Phosphate (PO₄):≤100 ppm
- EP/BP - Sulfate (SO₄):≤200 ppm
- EP/BP - Iron (Fe):≤10 ppm
- EP/BP - Sodium (Na):≤1.0 %
- FCC - Assay (total alkali as KOH):85.0 - 100.5 %
- FCC - Carbonate (as K₂CO₃):≤2.0 %
- FCC - Identification:Passes Test
- FCC - Insoluble Substances:Passes Test
- FCC - Lead (Pb):≤2 mg/kg
- FCC - Mercury (Hg)(mg/kg):≤0.1
- Solubility:Passes Test
- Endotoxin Concentration (EU/g):≤5.0
Specification Test Results
About this item
:
:56.11 g/mol
Merck Index:12,07806
Boiling Pt:1320 °C (1013 hPa)
Melting Pt:360 °C
Density:2,04 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-181-3
UN:1813
ADR:8,II
REACH:01-2119487136-33
:GHS05
:GHS07
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553