Potassium chromate ≥99.5%, crystals, AR® ACS, Macron Fine Chemicals™
Catalog # 6870-20
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pk:12 kg
- Seq:4187
- Environmentally Preferable:
Specifications
Specification Test Results
- Meets ACS Reagent Chemical Requirements:—
- For Laboratory,Research,or Manufacturing Use:= 99.5 %
- ACS - Assay (K₂CrO₄):≥ 99.5 %
- ACS - Calcium (Ca):≤ 0.005 %
- ACS - Chloride (Cl):≤ 0.005 %
- ACS - Insoluble Matter:≤ 0.005 %
- ACS - pH of 5% Solution at 25°C:8.6 - 9.8
- ACS - Sodium (Na):≤ 0.02 %
- ACS - Sulfate (SO₄):≤ 0.03 %
Specification Test Results
About this item
:
:194.19 g/mol
UN:3288
:GHS08
:GHS06
:GHS09
CAS Number:7789-00-6
Formula:K₂CrO₄
MDL Number:MFCD00011368
Boiling Pt:1000
Melting Pt:985 °C
ADR:6.1 III
Merck Index:12,07784