Order Entry
Ireland
ContactUsLinkComponent
Phenol USP, Macron Fine Chemicals™
Phenol USP, Macron Fine Chemicals™
Catalog # 0610-06
CAS Number:  
undefined
Phenol USP, Macron Fine Chemicals™
Catalog # 0610-06
CAS Number:  

Specifications

  • Pk:
    2,5 kg
  • Seq:
    4076

Specifications

Specification Test Results

  • Bulk Pharmaceutical Chemical:
    = 89.0 %
  • USP - Assay (C₆H₅OH):
    ≥89.0 %
  • USP - Benzene:
    ≤2.0 ppm
  • USP - Identification A:
    Passes Test
  • USP - Identification B:
    Passes Test
  • USP - Clarity of Solution and Reaction:
    Passes Test
  • USP - Nonvolatile Residue:
    ≤0.05 %
  • Distilling Range - Initial Temperature (For Information Only):
    = 182.5 °C
  • Distilling Range - Final Temperature:
    ≤182.5 °C
  • Stabilizer:
    Passes Test
  • JP - Identification (1):
    Passes Test
  • JP - Identification (2):
    Passes Test
  • JP - Boiling Point:
    ≤182 °C
  • JP - Clarity and Color of Solution and Acidity or Alkalinity:
    Passes Test
  • JP - Residue on Evaporation:
    ≤0.05 %
  • JP - Assay:
    ≥88.0 %

Specification Test Results

About this item

 :
 :94.11 g/mol
UN:1671
ADR:6.1,II
CAS Number:108-95-2
Formula:C₆H₅OH
MDL Number:MFCD00002143