Petroleum spirit, 30…60 °C, BAKER ANALYZED® for gas chromatography, J.T.Baker®
Catalog # 9200.2500
Supplier: Avantor
CAS Number:
Specifications
- Pk:2,5 L
- Seq:5096
- Environmentally Preferable:
Specifications
Specification Test Results
- Color (APHA):max. 10
- Residue after Evaporation:max. 5 ppm
- Water (H2O):max. 0.05 %
- Hydrocarbon oil index concentration (as RIVM oil reference standard):max. 0.5 mg/l
- Total peaks between n-decane (C10H22) and n-tetracontane (C40H82):max. 0.5 mg/l
Specification Test Results
About this item
Synonyms: Gasoline, Petroleum ether, Petrol, Benzine
:
UN:1268
ADR:3,II
:GHS02
:GHS07
:GHS08
:GHS09
CAS Number:64742-49-0
:64742-82-1
MDL Number:MFCD00081849