Anti-CGRP Rabbit Polyclonal Antibody
Catalog # BIRBORB182870-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Calcitonin Gene Related Peptide
- Antigen symbol:CGRP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Storage temperature:Store at 4 °C for up to two weeks. For long term storage, aliquot and store at –20 °C, avoid freeze/thaw cycles
- Immunogen:KLH conjugated synthetic peptide derived from human CGRP-I
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: CGRP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat