Order Entry
Ireland
ContactUsLinkComponent
Anti-MERTK (phospho-Tyr749) Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-MERTK (phospho-Tyr749) Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
  BIRBORB189891-100
 :  Biorbyt
Anti-MERTK (phospho-Tyr749) Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
  BIRBORB189891-100
 :  Biorbyt

 

  • Antibody type:
    Primary
  • Antigen symbol:
    MERTK (phospho-Tyr749)
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Storage temperature:
    Store at 4 °C for up to two weeks. For long term storage, aliquot and store at –20 °C, avoid freeze/thaw cycles
  • Immunogen:
    KLH conjugated synthesised phosphopeptide derived from human MERTK around the phosphorylation site of Tyr749
  • Pk:
    100 µG

 

 

Type: Primary
Antigen: MERTK (phospho-Tyr749)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat