Order Entry
Ireland
ContactUsLinkComponent
Chemical resistant shoe covers, MICROGARD® 2000, model 400
Chemical resistant shoe covers, MICROGARD® 2000, model 400
Catalog # ANSEWH20B-00400-00
Supplier:  Ansell
undefined
Chemical resistant shoe covers, MICROGARD® 2000, model 400
Catalog # ANSEWH20B-00400-00
Supplier:  Ansell
Specifications
  • Shoe size:
    42 - 46
  • Garment application:
    Laboratory
  • Colour:
    White
  • Seq:
    04
  • Pk:
    400 PAIR
Specifications
About this item
These over shoes are made from a microporous PE film with a non woven inner layer. This fabric provides low hazard liquid chemical repllence particle protection, protection from pesticides and a barrier to biological agents.
  • Elasticated opening
  • Bound seams
  • Fit shoe size 42 to 46