Bottles, narrow neck, alpha, lightweight with screw caps
Supplier: Azpack
Certificates
About this item
Clear or amber lightweight glass.
- For use in applications without high thermal loads
- Amber coloured to protect light-sensitive material
- A lighter, stronger and more uniform range of bottles
Large range of standard round glass bottles. All sizes of the lightweight bottles have exactly the same neck which means that only one cap size for the entire range has to be stocked.
Frequently Bought Together