Acid detergent descaler; RBS T 305
CHEPT305/E123
: CHEMICAL
- Description:Acid detergent descaler; RBS T 305
- Pk:5 L
- pH range:1.5 (2%)
Cleaning agent, descaler and oxides remover.
- Low use concentration
- High detergency power
- Contains phosphoric acid
RBS T 305 is suitable for manual cleaning, soaking and ultrasonic cleaning of items and laboratory equipment. It was especially designed to remove mineral residues such as calcareous deposits, rust, insoluble salts etc. It is particularly adapted for cleaning animal cages and rooms in veterinary institutions.
: Available in box of 4 × 5 litres. Please contact Avantor for details.