Order Entry
Ireland
ContactUsLinkComponent
Alkaline cleaning solution, liquid concentrate, Micro-90®
Alkaline cleaning solution, liquid concentrate, Micro-90®
  IPROM-9033
   Safer Human and Environmental Health
Alkaline cleaning solution, liquid concentrate, Micro-90®
  IPROM-9033
   Safer Human and Environmental Health
  • Description:
    Micro-90® alkaline cleaning solution
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Pk:
    5 gal US
Micro-90® is a mild, yet powerful, multipurpose, alkaline cleaning concentrate that has long been used in laboratories, industrial applications, and critical cleaning processes.
  • Manufactured in a dedicated system
  • pH: 9,5
Micro-90® is a unique chelating detergent that contains ionic and non-ionic ingredients, which combine to produce a variety of cleaning actions. It lifts, disperses, emulsifies, sequesters, suspends, and decomposes soils, then rinses away leaving the surface absolutely clean. Excellent for use on stainless steel, glass, ceramic, plastic, rubber, concrete, filter membranes and most other hard surfaces.
 : Registered with NSF as an A1 cleaner and can be validated in FDA processes.