Order Entry
Ireland
ContactUsLinkComponent
Azelaic acid ≥98% (by GC) for synthesis
Irritant
Azelaic acid ≥98% (by GC) for synthesis
Supplier:  Merck
Certificates
Irritant
Azelaic acid ≥98% (by GC) for synthesis
Supplier:  Merck

Specification Test Results

  • Assay (GC, area%):
    ≥98,0% (a/a)
  • Melting range (lower value):
    ≥106 °C
  • Melting range (upper value):
    ≤109 °C
  • Identity (IR):
    Passes test
  • Molar mass :
    188,22 g/mol
  • Vapor pressure:
    1,3 hPa (178,3 °C)
  • Solubility:
    2,4 g/l
  • Storage:
    Store below +30 °C

Specification Test Results

Related Information

Frequently Bought Together