Order Entry
Ireland
ContactUsLinkComponent
Dual-gel electrophoresis system, Owl™ P10DS
Dual-gel electrophoresis system, Owl™ P10DS
Catalog # 730-0263
undefined
Dual-gel electrophoresis system, Owl™ P10DS
Catalog # 730-0263
Specifications
  • Model:
    Owl P10DS
  • Description:
    Owl P10DS, complete system with gel caster
  • Pk:
    1
Specifications
About this item
The Owl™ P10DS dual gel system casts and runs one or two 20×20 cm gels in the upper buffer chamber. The larger size cassette allows the maximum number of samples and separation length. The system is ideal for second dimension protocols of 2-D electrophoresis applications.
  • Optimal for large sample numbers that need extended separation and mobility shift assays
Gel W×H: 200×200 mm
Footprint W×H×D: 310×230×165 mm
Running buffer volume: 1150 ml (upper chamber 650 ml, lower chamber 500 ml)
Delivery: Complete system includes upper buffer chamber with internal cooling core, lower buffer chamber, SuperSafe lid with attached power leads, 4 blank glass plates, 4 notched glass plates, 4 combs 1,5 mm thick (2×15 well, 2×20 well), spacer set (1,5 mm thick), spacer placer, and blocking plate for single gel operation.
Combs
Product Information