About this item
Knitted nylon gloves with nitrile foam dip coating on the palm.
- Dissipation of electrostatic discharge; anti-static according to EN 1149
- Thin liner for extra softness and comfort
- Hand-specific: Ergonomically designed for excellent dexterity, comfort and long wear
- No solvent-based materials, no silicone
- Glove’s light colour helps users easily detect any dirt or contamination
Especially designed for micro-electronic work where electro-static discharge (ESD) must be avoided. The thin design improves the handling of ever smaller components. Also suitable for use in pre and post painting areas and controlled-environment rooms.
Gauge: 13
Colour: Grey
EN 388 (2131), EN 1149, Cat. II
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Catalog No:
- 112-3194
- 112-3191
- 112-3193
- 112-3190
- 112-3192
- ANSE11-100/10
- Model:
- 11-100
- 11-100
- 11-100
- 11-100
- 11-100
- 11-100
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Nylon/conductive fibres
- Nylon/conductive fibres
- Nylon/conductive fibres
- Nylon/conductive fibres
- Nylon/conductive fibres
- Nylon/conductive fibres
- Finish:
- Palm coated
- Palm coated
- Palm coated
- Palm coated
- Palm coated
- Palm coated
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (2131A)
- EN 388:2016 (2131A)
- EN 388:2016 (2131A)
- EN 388:2016 (2131A)
- EN 388:2016 (2131A)
- EN 388:2016 (2131A)
- Anti-static:
- +
- +
- +
- +
- +
- +