Hiker socks, SK12
Supplier: Portwest
Certificates
About this item
These socks are made from 50% cotton, 30% Coolmax® fibre and 20% nylon. Ideal for work or leisure.
- Coolmax fabric for moisture management
- Ensure a dry environment for comfortable and healthy feet
- Reinforced heels and toes
- Cushion sole area
Specifications
- Catalog No:
- PRTWSK12BKR39-43
- PRTWSK12BKR44-48
- Model:
- SK12
- SK12
- Garment application:
- Casual, General Purpose
- Casual, General Purpose
- Gender:
- Unisex
- Unisex
- Fabric:
- 50% cotton, 30% Coolmax® fibre and 20% nylon
- 50% cotton, 30% Coolmax® fibre and 20% nylon
Specifications
Frequently Bought Together