Detergents, InSpec™ N10
DYCEINSPECN10
: DYCEM
- Description:InSpec™ N10 Solution
- Packed:6×1 l trigger spray
- Pk:6
Inspec N10 is a powerful non sterile solution of mildly alkaline detergent specifically used for cleanroom walls, floors, ceilings and work surfaces. It is most effective at removing general process derived residue and soiling.
- Rapid action alkaline detergent
- Suitable for manual cleaning
- Defined 24 month shelf life
This is also available as a pre-diluted detergent, made with WFI quality water.Available in sterile or non sterile preparations.
: Manufactured to GMP standards in an ISO 6 cleanroom, filled and capped in an ISO class 5 environment.
: Solutions are filled through a 0,2 micron filter into final packaging.