Specifications
- Antibody type:Primary
- Antigen name:Leucine-Rich Repeats And Calponin Homology (CH) Domain Containing 3
- Antigen symbol:LRCH3
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:84859
- Antigen synonyms:LRCH3_HUMAN.|LRCH3|MGC4126|Leucine rich repeats and calponin homology CH domain containing 3
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Members of the leucine-rich repeat family includes LRCH1, LRCH2, LRCH3 and LRCH4. All family members contain one calponin-homology domain and nine leucine-rich repeats. The best characterized leucine-rich repeat family member is LRCH4, which is suggested to be involved in ligand binding in the brain, with expression observed primarily in the hippocampus. As a cell adhesion molecule and signal receptor, LRCH4 may play an important role in maintenance of hippocampus-dependent memories, with defects in the gene possibly contributing to a loss of long-term memory. The gene encoding LRCH3 maps to human chromosome 3, which spans 200 million base pairs and encodes between 1,100 and 1,500 genes. There are three isoforms of LRCH3 that are produced as a result of alternative splicing events.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: LRCH3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat