Anti-HRNR Rabbit Polyclonal Antibody (Cy5.5®)
Catalog # BOSSBS-9703R-CY5.5
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:hornerin
- Antigen symbol:HRNR
- Clonality:Polyclonal
- Conjugation:Cy5.5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Cross adsorption:No
- Form:liquid
- Gene ID:388697
- Antigen synonyms:S100A16|FLG3|S100a18
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Hornerin has similar structural features to those of profilaggrin, an essential protein for keratinization of epidermal tissues. Hornerin may play a role in cornification.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: HRNR
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human