Anti-AMPD3 Rabbit Polyclonal Antibody (Cy7®)
BOSSBS-9517R-CY7
: Bioss
- Antibody type:Primary
- Antigen name:AMP deaminase 3
- Antigen symbol:AMPD3
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:272
- Antigen synonyms:AMP deaminase 3|Erythrocyte AMP deaminase|AMPD3|AMP deaminase isoform E
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
AMP deaminase plays a critical role in energy metabolism.Involvement in diseaseDefects in AMPD3 are the cause of adenosine monophosphate deaminase deficiency erythrocyte type (AMPDDE); also known as erythrocyte AMP deaminase deficiency. AMPDDE is a metabolic disorder due to lack of activity of the erythrocyte isoform of AMP deaminase. It is a clinically asymptomatic condition characterized by a 50% increase in steady-state levels of ATP in affected cells. Individuals with complete deficiency of erythrocyte AMP deaminase are healthy and have no hematologic disorders.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: AMPD3
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat