Anti-GTF2H1 Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-8175R-CY7
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:General Transcription Factor 2H1
- Antigen symbol:GTF2H1
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:2965
- Antigen synonyms:Basic transcription factor 2 62 kDa subunit|P62|BTF2|Basic transcription factor 62 kDa subunit|TFB1|BTF2 p62|General transcription factor IIH polypeptide 1|gtf2h1|General transcription factor IIH subunit 1|BTF 2|TFIIH|TFIIH basal transcription factor complex p62 subunit|General transcription factor IIH,polypeptide 1,62kDa|TF2H1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:230-280/548
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Component of the core-TFIIH basal transcription factor involved in nucleotide excision repair (NER) of DNA and, when complexed to CAK, in RNA transcription by RNA polymerase II.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: GTF2H1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat