Anti-CCDC80 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-7992R-FITC
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:coiled-coil Domain Containing 80
- Antigen symbol:CCDC80
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:151887
- Antigen synonyms:DRO1|okuribin|SSG1|URB
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Promotes cell adhesion and matrix assembly.Tissue specificity: Expressed in dermal papilla and dermal fibroblasts (at protein level). Expressed in heart, thymus, placenta, pancreas, colon, epithelium, spleen and osteoblasts.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CCDC80
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat