Anti-C1QTNF1 Rabbit Polyclonal Antibody (Cy7®)
BOSSBS-7528R-CY7
: Bioss
- Antibody type:Primary
- Antigen name:C1q And tumor necrosis factor related protein 1
- Antigen symbol:C1QTNF1
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:114897
- Antigen synonyms:ZSIG37|CTRP1|GIP
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at –20 °C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
CTRP1 enhances the production of aldosterone. CTRP1 was highly expressed in obese subjects as well as up-regulated in hypertensive patients, CTRP1 may be a newly identified molecular link between obesity and hypertension.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: C1QTNF1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat