Anti-BRF1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
BOSSBS-6358R-FITC
: Bioss
- Antibody type:Primary
- Antigen name:BRF1, RNA polymerase III transcription initiation factor 90 kDa subunit
- Antigen symbol:BRF1
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:2972
- Antigen synonyms:HEL-S-76p|TFIIIB90|TAF3B2|TAFIII90|GTF3B|hBRF|RF|TF3B90|RNA polymerase III transcription initiation factor 90 kDa subunit|TAF3C|BRF-1|CFDS
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
General activator of RNA polymerase which utilizes different TFIIIB complexes at structurally distinct promoters. The isoform 1 is involved in the transcription of tRNA, adenovirus VA1, 7SL and 5S RNA. Isoform 2 is required for transcription of the U6 promoter.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: BRF1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat