Anti-CYP2W1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-6277R-A647
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Cytochrome P450, Family 2, Subfamily W, Polypeptide 1
- Antigen symbol:CYP2W1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:54905
- Antigen synonyms:Cytochrome P450|CYPIIW1|CYP2W1|CYP2W 1|MGC34287.|polypeptide 1|CP2W1_HUMAN|family 2|Cytochrome P450 2W1|subfamily W
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CYP2W1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype:
Reactivity: