Anti-RASAL Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-6088R-A488
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:RAS Protein Activator Like 1
- Antigen symbol:RASAL
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:8437
- Antigen synonyms:RAS protein activator like 1 (GAP1 like)|GAP1 like protein|RASAL|RAS-GTPase-activating protein-like|RAS protein activator like 1|RASAL1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:460-510/805
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Probable inhibitory regulator of the Ras-cyclic AMP pathway.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: RASAL
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat