Anti-ARMCX3 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Catalog # BOSSBS-5990R-A555
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Armadillo Repeat Containing, X-linked 3
- Antigen symbol:ARMCX3
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:51566
- Antigen synonyms:Arm protein lost in epithelial cancers|dJ545K15.2|GASP6|ARM protein lost in epithelial cancers on chromosome X 3|Armadillo repeat containing X linked protein 3|X chromosome|ALEX3|3
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
This gene encodes a member of the ALEX family of proteins which may play a role in tumor suppression. The encoded protein contains a potential N-terminal transmembrane domain and a single Armadillo (arm) repeat. Other proteins containing the arm repeat are involved in development, maintenance of tissue integrity, and tumorigenesis. This gene is closely localized with other family members on the X chromosome. Three transcript variants encoding the same protein have been identified for this gene. [provided by RefSeq, Jul 2008].
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ARMCX3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat