Anti-IL7R Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-5395R-A350
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Interleukin 7 Receptor
- Antigen symbol:IL7R
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:3575
- Antigen synonyms:IL7RA|CD127|ILRA|CDW127|IL-7R-alpha
- Modification:phospho-Tyr449
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:440-451/459
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Receptor for interleukin-7. Also acts as a receptor for thymic stromal lymphopoietin (TSLP).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: IL7R
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat