Order Entry
Ireland
ContactUsLinkComponent
Anti-G6PDH Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-G6PDH Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
  BOSSBS-4974R-FITC
 :  Bioss
Anti-G6PDH Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
  BOSSBS-4974R-FITC
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Glucose 6 Phosphate Dehydrogenase
  • Antigen symbol:
    G6PDH
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Antigen synonyms:
    G6PD1|Glucose-6-phosphate 1-dehydrogenase|G6pdx|G6PD|G6PD_HUMAN|Glucose 6 phosphate 1 dehydrogenase|POS10|Zwf1p.|MET19
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

Defects in G6PD are the cause of chronic non-spherocytic hemolytic anemia (CNSHA) . Deficiency of G6PD is associated with hemolytic anemia in two different situations. First, in areas in which malaria has been endemic, G6PD-deficiency alleles have reached high frequencies (1% to 50%) and deficient individuals, though essentially asymptomatic in the steady state, have a high risk of acute hemolytic attacks. Secondly, sporadic cases of G6PD deficiency occur at a very low frequencies, and they usually present a more severe phenotype. Several types of CNSHA are recognized. Class-I variants are associated with severe NSHA; class-II have an activity <10% of normal; class-III have an activity of 10% to 60% of normal; class-IV have near normal activity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Glucose 6 Phosphate Dehydrogenase
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat