Anti-TNNI Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-3458R-A647
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Troponin I
- Antigen symbol:TNNI
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:7138
- Modification:phospho-Ser23/Ser24
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:1-50/210
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Troponin T is the tropomyosin-binding subunit of troponin, the thin filament regulatory complex which confers calcium-sensitivity to striated muscle actomyosin ATPase activity.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: Troponin I
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human