Order Entry
Ireland
ContactUsLinkComponent
Anti-PRKAG3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Anti-PRKAG3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-3968R-A350
Supplier:  Bioss
Anti-PRKAG3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-3968R-A350
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Protein Kinase, AMP-activated, Non-catalytic Subunit Gamma 3
  • Antigen symbol:
    PRKAG3
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 350
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    53632
  • Antigen synonyms:
    AMPKG3|AMPK gamma3|gamma-3 subunit|AMPK gamma-3 chain|5'-AMP-activated protein kinase
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    174-220/489
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

AMP/ATP-binding subunit of AMP-activated protein kinase (AMPK), an energy sensor protein kinase that plays a key role in regulating cellular energy metabolism. In response to reduction of intracellular ATP levels, AMPK activates energy-producing pathways and inhibits energy-consuming processes: inhibits protein, carbohydrate and lipid biosynthesis, as well as cell growth and proliferation. AMPK acts via direct phosphorylation of metabolic enzymes, and by longer-term effects via phosphorylation of transcription regulators. Also acts as a regulator of cellular polarity by remodeling the actin cytoskeleton; probably by indirectly activating myosin. Gamma non-catalytic subunit mediates binding to AMP, ADP and ATP, leading to activate or inhibit AMPK: AMP-binding results in allosteric activation of alpha catalytic subunit (PRKAA1 or PRKAA2) both by inducing phosphorylation and preventing dephosphorylation of catalytic subunits. ADP also stimulates phosphorylation, without stimulating already phosphorylated catalytic subunit. ATP promotes dephosphorylation of catalytic subunit, rendering the AMPK enzyme inactive.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PRKAG3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat