Anti-LIPG Rabbit Polyclonal Antibody (Alexa Fluor® 647)
BOSSBS-2397R-A647
: Bioss
- Antibody type:Primary
- Antigen name:Lipase endothelial
- Antigen symbol:LIPG
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:9388
- Antigen synonyms:EDL|EL|PRO719
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:310-360/500
- Purification:Purified by Protein A
- Pk:100 µl
Has phospholipase and triglyceride lipase activities. Hydrolyzes high density lipoproteins (HDL) more efficiently than other lipoproteins. Binds heparin.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: LIPG
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat