Anti-PDEF/SPDEF Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Catalog # BOSSBS-1866R-A555
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen symbol:PDEF/SPDEF
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:25803
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
SPDEF, also known as PDEF is an ETS transcription factor expressed in the prostate epithelial cells. It is thought to act as an androgen-independent transactivator of the prostate-specific antigen (PSA) promoter and also as a transcriptional activator for the SERPINB5 promoter. It is highly expressed in prostate carcinoma cells and is thought to be involved in the regulation of the prostate gland and/or prostate cancer development. It is also expressed in epithelial breast carcinoma.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PDEF/SPDEF
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat