Anti-DUSP1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
BOSSBS-1851R-A488
: Bioss
- Antibody type:Primary
- Antigen name:Dual Specificity Protein Phosphatase 1
- Antigen symbol:DUSP1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:1843
- Antigen synonyms:MAP kinase phosphatase-1|CL100|PTPN10|Mitogen-activated protein kinase phosphatase 2|MKP-1|Dual specificity protein phosphatase hVH1|MKP II|DUSP 4|Dual specificity protein phosphatase hVH2|DUSP-4|VH1|MAP kinase phosphatase 2|Dual specificity protein phosphatase 1|Protein-tyrosine phosphatase CL100|MKP1|MKP-2|MKPII
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:175-225/367
- Purification:Purified by Protein A
- Pk:100 µl
Dual specificity phosphatase that dephosphorylates MAP kinase MAPK1/ERK2 on both 'Thr-183' and 'Tyr-185', regulating its activity during the meiotic cell cycle.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: DUSP1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat