Order Entry
Ireland
ContactUsLinkComponent
 
Anti-TROP2 Rabbit Polyclonal Antibody (Cy3®)
Catalog # BOSSBS-1243R-CY3
Supplier:  Bioss
Anti-TROP2 Rabbit Polyclonal Antibody (Cy3®)
Catalog # BOSSBS-1243R-CY3
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Tumor-associated calcium signal transducer 2
  • Antigen symbol:
    TROP2
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy3®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    7169
  • Antigen synonyms:
    Tumor-associated calcium signal transducer 2|Cell surface glycoprotein Trop-2|M1S1|GA733-1|TACSTD2|TROP2|Pancreatic carcinoma marker protein GA733-1
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    10-60/284
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

Binds to actin filaments in muscle and non-muscle cells. Plays a central role, in association with the troponin complex, in the calcium dependent regulation of vertebrate striated muscle contraction. Smooth muscle contraction is regulated by interaction with caldesmon. In non-muscle cells is implicated in stabilizing cytoskeleton actin filaments. The non-muscle isoform may have a role in agonist-mediated receptor internalization (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TROP2
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat