Anti-YWHAQ Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Catalog # BOSSBS-12419R-A350
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, theta
- Antigen symbol:YWHAQ
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:10971
- Antigen synonyms:14-3-3 protein T-cell|14-3-3 protein tau|14-3-3|HS1|protein tau|14-3-3 protein theta|1C5
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Adapter protein implicated in the regulation of a large spectrum of both general and specialized signaling pathways. Binds to a large number of partners, usually by recognition of a phosphoserine or phosphothreonine motif. Binding generally results in the modulation of the activity of the binding partner. Negatively regulates the kinase activity of PDPK1.This antibody is reactive with 14-3-3 Alpha/Beta/Gamma/Delta/Epsilon.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: YWHAQ
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat