Order Entry
Ireland
ContactUsLinkComponent
Anti-P2RY8 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anti-P2RY8 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-12073R-FITC
Supplier:  Bioss
Anti-P2RY8 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-12073R-FITC
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Purinergic Receptor P2Y, G-Protein Coupled 8
  • Antigen symbol:
    P2RY8
  • Clonality:
    Polyclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Form:
    liquid
  • Antigen synonyms:
    RP11-261P4.4|MGC50878|P2Y8|G-Protein Coupled Purinergic Receptor P2Y8
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

G protein-coupled receptors (GPRs), also known as seven transmembrane receptors, heptahelical receptors or 7TM receptors, comprise a superfamily of proteins that play a role in many different stimulus-response pathways. G-protein coupled receptors translate extracellular signals into intracellular signals (G-protein activation) and they respond to a variety of signaling molecules, such as hormones and neurotransmitters. P2RY8 (purinergic receptor P2Y, G-protein coupled, 8), also known as P2Y8, is a 359 amino acid multi-pass membrane protein that localizes to the cell membrane and belongs to the G protein-coupled receptor family. Expressed at low levels in lung, heart and kidney, P2RY8 may function as a receptor for purines that are coupled to G proteins and may also play a role in mental retardation.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: P2RY8
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human