Order Entry
Ireland
ContactUsLinkComponent
Human IgA Isotype Control
Human IgA Isotype Control
Catalog # ICNA0855906
Supplier:  MP Biomedicals
Human IgA Isotype Control
Catalog # ICNA0855906
Supplier:  MP Biomedicals

Specifications

  • Antibody type:
    Control
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Human
  • Isotype:
    IgA
  • Reactivity:
  • Environmentally Preferable:
  • Form:
    Lyophilized
  • Concentration:
    Total Protein: 5.41 mg/mL; Volume: 1 mL
  • Shipping temperature:
    RT
  • Purification:
    Human IgA is purified from pooled human serum using multi-step procedures which may include salt fractionation, gel filtration, ion-exchange chromatography and immunoabsorption. The product is dialyzed into 0.01M sodium phosphate, 0.07M sodium chloride, pH 7.3, filtered through a 0.22 μm filter, vialed and lyophilized.
  • Size:
    5 mg
  • Pk:
    5 mg

Specifications

About this item

Product is the lyophilized powder of human serum IgA and buffer salts. The product is tested for purity at a minimum of 20 mg/mL using immunoelectrophoresis. The product is human serum IgA; no trace of IgG, IgM or non-immunoglobulin serum proteins is detected. The total protein is determined using the Biuret procedure, with bovine albumin as the standard. Each vial is overfilled by approximately 10 percent so that a minimum of 5 mg of human serum IgA is obtained upon reconstitution.