Anti-C/EBP-beta Rabbit Polyclonal Antibody [clone: S229/182]
Catalog # BWRLBS1037
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:C/EBP- beta
- Clonality:Polyclonal
- Clone:S229/182
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 200-250 of Human C/EBP-beta.
C/EBP-? (S229/182) pAb detects endogenous levels of total C/EBP-? protein.
Type: Primary
Antigen: C/EBP-beta
Clonality: Polyclonal
Clone: S229/182
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat