Anti-Thrombin R Rabbit Polyclonal Antibody [clone: S42]
Catalog # BWRLBS7067
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Thrombin R
- Clonality:Polyclonal
- Clone:S42
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 12-60 of Human Thrombin R.
Thrombin R (S42) pAb detects endogenous levels of Thrombin R protein.
Type: Primary
Antigen: Thrombin R
Clonality: Polyclonal
Clone: S42
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human