Anti-TXNRD2 Mouse Monoclonal Antibody
BWRLMB0170
- Antibody type:Primary
- Antigen symbol:TXNRD2
- Clonality:Monoclonal
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Recombinant full length Human TXNRD2.
This antibody detects endogenous levels of TXNRD2 and does not cross-react with related proteins
Type: Primary
Antigen: TXNRD2
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human