Anti-Dynactin 1 Mouse Monoclonal Antibody
Catalog # BWRLMB0011
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Dynactin 1
- Clonality:Monoclonal
- Host:Mouse
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to the N-terminal of Human Dynactin 1.
This antibody detects endogenous levels of DCTN1 and does not cross-react with related proteins
Type: Primary
Antigen: Dynactin 1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope: N-terminal
Host: Mouse
Isotype:
Reactivity: Human