Anti-HNRNPU Mouse Monoclonal Antibody
BWRLMB0002
- Antibody type:Primary
- Antigen symbol:HNRNPU
- Clonality:Monoclonal
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Recombinant full length Human HNRNPU.
This antibody detects endogenous levels of HNRNPU and does not cross-react with related proteins
Type: Primary
Antigen: HNRNPU
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human