Anti-IL-18R Rabbit Polyclonal Antibody [clone: M270]
Catalog # BWRLBS9268
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:IL-18R
- Clonality:Polyclonal
- Clone:M270
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 236-288 of Human IL-18R.
IL-18R (M270) pAb detects endogenous levels of IL-18R protein.
Type: Primary
Antigen: IL-18R
Clonality: Polyclonal
Clone: M270
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human