Anti-NEDD9 Rabbit Polyclonal Antibody
Catalog # BWRLBS7372
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:NEDD9
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recombinant full length Human NEDD9.
NEDD9 pAb detects endogenous levels of NEDD9 protein.
Type: Primary
Antigen: NEDD9
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat