Anti-PSMC4 Rabbit Polyclonal Antibody
BWRLBS7365
- Antibody type:Primary
- Antigen symbol:PSMC4
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Recombinant full length Human PSMC4.
PSMC4 pAb detects endogenous levels of PSMC4 protein.
Type: Primary
Antigen: PSMC4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat