Order Entry
Ireland
ContactUsLinkComponent
 
Anti-PSMA2 Rabbit Polyclonal Antibody
  BWRLBS7364
Anti-PSMA2 Rabbit Polyclonal Antibody
  BWRLBS7364

 

  • Antibody type:
    Primary
  • Antigen symbol:
    PSMA2
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Recombinant full length Human PSMA2.

PSMA2 pAb detects endogenous levels of PSMA2 protein.

Type: Primary
Antigen: PSMA2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat