Anti-FOXM1 Rabbit Polyclonal Antibody
Catalog # BWRLBS7358
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:FOXM1
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recombinant full length Human FOXM1.
FOXM1 pAb detects endogenous levels of FOXM1 protein.
Type: Primary
Antigen: FOXM1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat