Anti-UGT1A1 Rabbit Polyclonal Antibody
BWRLBS7952
- Antibody type:Primary
- Antigen symbol:UGT1A1
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Recombinant full length Human UGT1A1.
UGT1A1 pAb detects endogenous levels of UGT1A1 protein.
Type: Primary
Antigen: UGT1A1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human